Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555546
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555546 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555546 Supplier Invitrogen™ Supplier No. PA555546
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEPVVYQKLL PWEPSLESEE EVEEEETSEA LVLNPRRHQD SSRNKAGGLP GTWVRVVAAL LLLAVGCSLA VRQLQNQGRS Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 59%.

Cleaves glutathione conjugates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGT6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GGT6
Gene Accession No. Q6P531
Gene Alias 9030405D14Rik; AI427679; AW490214; gamma-glutamyl transferase 6; gamma-glutamyltransferase 6; gamma-glutamyltransferase 6 homolog; gamma-glutamyltranspeptidase 6; GGT 6; GGT6; Glutathione hydrolase 6; Glutathione hydrolase 6 heavy chain; Glutathione hydrolase 6 light chain
Gene Symbols GGT6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GGT6. Recombinant protein control fragment (Product #RP-93386).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124975
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.