Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GGT7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553035
Description
Immunogen sequence: HLVLSPPPPH TGPALISALN ILEGFNLTSL VSREQALHWV AETLKIALAL ASRLGDPVYD STITESMDDM LSKVEAAYLR GHINDSQAAP APLLPVYELD GAPTAAQVLI MGPDDFIVAM VSSLNQPFGS GLITPSGILL NS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
GGTL3 is an enzymes involved in both the metabolism of glutathione and in the transpeptidation of amino acids. Changes in the activity of gamma-glutamyltransferase may signal preneoplastic or toxic conditions in the liver or kidney. GGTL3 consists of a heavy and a light chain, and it can interact with CT120, a plasma membrane-associated protein that is possibly involved in lung carcinogenesis.This gene is a member of a gene family that encodes enzymes involved in both the metabolism of glutathione and in the transpeptidation of amino acids. Changes in the activity of gamma-glutamyltransferase may signal preneoplastic or toxic conditions in the liver or kidney. The protein encoded by this gene consists of a heavy and a light chain, and it can interact with CT120, a plasma membrane-associated protein that is possibly involved in lung carcinogenesis.
Specifications
GGT7 | |
Polyclonal | |
Unconjugated | |
Ggt7 | |
1110017C11Rik; 6330563L03Rik; D20S101; dJ18C9.2; gamma-glutamyltransferase 4; gamma-glutamyltransferase 7; gamma-glutamyltransferase 7; glutathione hydrolase 7; gamma-glutamyltransferase-like 3; Gamma-glutamyltransferase-like 5; gamma-glutamyltranspeptidase 7; gamma-glutamyltranspeptidase-like 3; GGT 7; GGT4; Ggt7; GGTL3; GGTL5; Glutathione hydrolase 7; Glutathione hydrolase 7 heavy chain; Glutathione hydrolase 7 light chain; LOC100732094; mFLJ00311 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
156275, 207182, 2686 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99JP7, Q99MZ4, Q9UJ14 | |
Ggt7 | |
Recombinant protein corresponding to Human GGT7. Recombinant protein control fragment (Product #RP-90211). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction