Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GGTI 298
GREENER_CHOICESDP

Catalog No. 24301 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-301 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 24-301 Supplier Tocris Bioscience™ Supplier No. 2430/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Geranylgeranyltransferase I (GGTase I) inhibitor

CAAZ peptidomimetic geranylgeranyltransferase I (GGTase I) inhibitor. Strongly inhibits the processing of geranylgeranylated Rap1A with little effect on processing of farnesylated Ha-Ras (IC50 values are 3 and > 10 μM respectively). Causes G0-G1 cell cycle block and apoptosis in A549 cells and inhibits cell invasion and migration in COLO 320CM cells.

Chemical Identifiers

CAS 1217457-86-7
Molecular Formula C29H34F3N3O5S

Specifications

Quantity 1 mg
Formula Weight 593.66
Purity >95%
Molecular Weight (g/mol) 593.662
Chemical Name or Material GGTI 298
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.