Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GGTLC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569148
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569148 Supplier Thermo Scientific Supplier No. PA569148
Add to Cart
Add to Cart

GGTLC1 Polyclonal Antibody

GGTLC1 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen GGTLC1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GGTLC1
Gene Accession No. Q9BX51
Gene Alias dJ831C21.1; dJ831C21.2; gamma-glutamyl transpeptidase; gamma-glutamyltransferase light chain 1; gamma-glutamyltransferase-like activity 3; gamma-glutamyltransferase-like activity 4; Gamma-glutamyltransferase-like protein 6; GGTL6; GGTLA3; GGTLA4; GGTLC1; Glutathione hydrolase light chain 1
Gene Symbols GGTLC1
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human GGTLA4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92086
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.