Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGTLC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523722
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523722 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523722 Supplier Invitrogen™ Supplier No. PA523722
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse, porcine and rat based on sequence homology.

Gamma-glutamyltransferase-1 (GGT1; MIM 612346) is a membrane-bound extracellular enzyme that cleaves gamma-glutamyl peptide bonds in glutathione and other peptides and transfers the gamma-glutamyl moiety to acceptors. Autocatalytic cleavage of the GGT1 precursor polypeptide produces a heavy chain and a light chain that associate with each other to form the functional enzyme. Lightchain-only GGTs, such as GGTLC2, contain a region corresponding to the GGT1 light chain, but they lack the membrane-anchoring heavy chain region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGTLC2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GGTLC2
Gene Accession No. Q14390
Gene Alias Gamma-glutamyltransferase light chain 2; gamma-glutamyltransferase-like 4; gamma-glutamyltransferase-like protein 4; GGTL4; GGTLC2; Glutathione hydrolase light chain 2
Gene Symbols GGTLC2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 127-154 amino acids from the central region of human GGTLC2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91227
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.