Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GH2/Growth hormone 2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982831
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2831 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2831 Supplier Bioss Supplier No. BS13354R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Plays an important role in growth control. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. It stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake and protein synthesis in muscle and other tissues.

Specifications

Antigen GH2/Growth hormone 2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GH2
Gene Accession No. P01242
Gene Alias GH2; GHB2; GHL; GHV; GH-V; Growth hormone 2; growth hormone B2; Growth hormone variant; hGH-V; placental-specific growth hormone; Placenta-specific growth hormone
Gene Symbols GH2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GH2/Growth hormone 2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2689
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.