Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GH2 Polyclonal Antibody

Rabbit Polyclonal Antibody

$209.50 - $511.50

Specifications

Antigen GH2
Concentration 500 μg/mL
Content And Storage -20°C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5119166
View Documents
Invitrogen™
PA5119166
200 μL
Each for $511.50
Only null left
Add to Cart
 
PIPA5121959
View Documents
Invitrogen™
PA5121959
20 μL
Each for $209.50
Only null left
Add to Cart
 
Description

Description

GH2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Plays an important role in growth control. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. It stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake and protein synthesis in muscle and other tissues.
Specifications

Specifications

GH2
-20°C
Polyclonal
Liquid
IgG
Human
GH2
GH2; GHB2; GHL; GHV; GH-V; Growth hormone 2; growth hormone B2; growth hormone variant; hGH-V; placental-specific growth hormone; Placenta-specific growth hormone
GH2
Primary
Antigen affinity chromatography
500 μg/mL
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300
P01242
2689
Recombinant protein Growth Hormone 2. The antigen corresponds to amino acid range 27-217 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.