Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GHDC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534464
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534464 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534464 Supplier Invitrogen™ Supplier No. PA534464
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is 293 cell lysate. PA5-34464 can be used with blocking peptide PEP-1506.

GHDC, also known as LGP1, is a GH3 domain-containing protein that is highly expressed in normal mammary tissues and mammary tumors. GHDC is primarily located in the nuclear envelope. In mouse, its gene is located on chromosome 11 adjacent to the STAT3/5 locus. Little is known of the function of this protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GHDC
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Ghdc
Gene Accession No. Q8N2G8, Q99J23
Gene Alias D11LGP1; D11Lgp1e; DL11LGP1; GH3 domain containing; GH3 domain-containing protein; Ghdc; GHDC1; homolog of mouse LGP1; LGP1; RGD1308881
Gene Symbols Ghdc
Host Species Rabbit
Immunogen A 19 amino acid peptide near the amino terminus of human GHDC.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303542, 80860, 84514
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.