Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GHDC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556808
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556808 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556808 Supplier Invitrogen™ Supplier No. PA556808
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SAPHYEVFVA LRGLRNLSEE NRDKLDHCLQ EASPRYKSLR FWGSVGPARV HLVGQGAFRA LRAALAACPS SPFPPAMPRV LRHRHLAQCL Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 86%.

GHDC, also known as LGP1, is a GH3 domain-containing protein that is highly expressed in normal mammary tissues and mammary tumors. GHDC is primarily located in the nuclear envelope. In mouse, its gene is located on chromosome 11 adjacent to the STAT3/5 locus. Little is known of the function of this protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GHDC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ghdc
Gene Accession No. Q8N2G8
Gene Alias D11LGP1; D11Lgp1e; DL11LGP1; GH3 domain containing; GH3 domain-containing protein; Ghdc; GHDC1; homolog of mouse LGP1; LGP1; RGD1308881
Gene Symbols Ghdc
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GHDC. Recombinant protein control fragment (Product #RP-93392).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84514
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.