Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GHITM Antibody (42), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP312773AF488
Description
GHITM Monoclonal antibody specifically detects GHITM in Invertebrate samples. It is validated for ELISASpecifications
GHITM | |
Monoclonal | |
Alexa Fluor 488 | |
Dermal papilla-derived protein 2, DERP2MICS1, DKFZp566C0746, FLJ26584, growth hormone inducible transmembrane protein, growth hormone-inducible transmembrane protein, HSPC282, Mitochondrial morphology and cristae structure 1, PTD010, TMBIM5My021, transmembrane BAX inhibitor motif containing 5, Transmembrane BAX inhibitor motif-containing protein 5 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dermatophagoides pteronyssinus GHITM (Uniprot#: P49278; Gln19-Asp146). | |
100 μg | |
Primary | |
Invertebrate | |
Purified |
ELISA | |
42 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
27069 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction