Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GHITM Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP147217
Description
GHITM Polyclonal specifically detects GHITM in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
GHITM | |
Polyclonal | |
Western Blot 0.01 - 0.1 ug/ml, Peptide ELISA Detection limit 1:8000 | |
NP_055209.2 | |
GHITM | |
Peptide with sequence C-PSREYATKTRIGIR corresponding to internal region according to NP_055209.2. | |
0.1 mg | |
Primary | |
This antibody recognizes and epitope within the internal region. Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog, Cow and Pig. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
Dermal papilla-derived protein 2, DERP2MICS1, DKFZp566C0746, FLJ26584, growth hormone inducible transmembrane protein, growth hormone-inducible transmembrane protein, HSPC282, Mitochondrial morphology and cristae structure 1, PTD010, TMBIM5My021, transmembrane BAX inhibitor motif containing 5, Transmembrane BAX inhibitor motif-containing protein 5 | |
Goat | |
Affinity Purified | |
RUO | |
27069 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction