Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ghrelin Monoclonal Antibody (121)
GREENER_CHOICE

Catalog No. ABS05212102
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS05212102 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS05212102 Supplier Invitrogen™ Supplier No. ABS05212102
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specificity: ABS 052-121-02 cross-reacts with rat ghrelin, and is predicted to react with mouse, dog and cat based on sequence homology. Reactivity: ABS 052-121-02 binds human ghrelin in solution. ABS 052-121-02 can be used as a detection antibody in a sandwich ELISA with capture antibody ABS 050-45-02. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

GHRL encodes ghrelin-obestatin preproprotein, which generates ghrelin and obestatin. Ghrelin is an the ligand for the growth hormone secretagogue receptor type 1 and is involved in regulating growth hormone release. Ghrelin has an appeptite-stimulating effect and induces adiposity and stimulates gastric acid secretion. Obestatin may be the ligand for the orphan G protein-coupled receptor GPR39 and is involved in satiety and can have an appetite-reducing effect resulting in decreased food intake.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ghrelin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 121
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene GHRL
Gene Accession No. Q6BEG6, Q9BEF8, Q9EQX0, Q9QYH7, Q9UBU3
Gene Alias 2210006E23Rik; AMCBX1; Appetite-regulating hormone; appetite-regulating hormone; uncharacterized protein LOC58991; exon 2- and 3-deleted preproghrelin; exon 2-deleted preproghrelin; FIP3; FIP-3; Fip3p; Ghr; Ghrelin; ghrelin 27; ghrelin 28; ghrelin and obestatin prepropeptide; ghrelin precursor; ghrelin, growth hormone secretagogue receptor ligand; ghrelin/obestatin; ghrelin/obestatin preprohormone; ghrelin/obestatin prepropeptide; Ghrelin-27; Ghrelin-28; ghrelin-like protein; GHRL; Growth hormone secretagogue; growth hormone-releasing peptide; IKK-gamma; In2c-preproghrelin; IP; IP1; IP2; IPD2; m46; Motilin-related peptide; MTLRP; MTLRPAP; NEMO; Obestatin; Obestatin-13; Obestatin-23; OTTHUMP00000207794; OTTHUMP00000207795; OTTHUMP00000207796; OTTHUMP00000207798; prepro-appetite regulatory hormone; preproghrelin; proghrelin; Protein M46; UNQ524/PRO1066
Gene Symbols GHRL
Host Species Mouse
Immunogen Synthetic ghrelin (14-28).
Purification Method Protein A/G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 403587, 493844, 51738, 58991, 59301
Target Species Human, Mouse, Rat, Canine, Feline
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.