Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ghrelin/Obestatin Antibody (45), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147183AF405
Description
Ghrelin/Obestatin Monoclonal specifically detects Ghrelin/Obestatin in Human samples. It is validated for Western Blot, ELISA.Specifications
Ghrelin/Obestatin | |
Monoclonal | |
Alexa Fluor 405 | |
50 mM sodium borate with 0.05% sodium azide | |
GHRL | |
Synthetic human Ser3-octanoyl ghrelin-28 | |
0.1 mL | |
51738.0 | |
Human | |
IgG1 λ |
Western Blot, ELISA | |
45 | |
Western Blot, ELISA | |
appetite-regulating hormone, ghrelin, ghrelin/obestatin preprohormone, ghrelin/obestatin prepropeptide, Growth hormone secretagogue, Growth hormone-releasing peptide, Motilin-related peptide, MTLRPgrowth hormone secretagogue receptor ligand, obestatin, Protein M46 | |
Mouse | |
Protein A or G purified | |
Primary | |
binds human ghrelin and human des-acyl ghrelin, cross-reacting ∼5% with rat ghrelin in sandwich ELISA with antibody as biotinylated detection antibody. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction