Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GHRH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592408 Supplier Invitrogen™ Supplier No. PA592408
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Raji, HL-60, U-937, Mouse liver.

The activated kinase acts on a variety of targets. Likely to be the GTPase effector that links the Rho-related GTPases to the JNK MAP kinase pathway. Activated by CDC42 and RAC1. Involved in dissolution of stress fibers and reorganization of focal complexes. Involved in regulation of microtubule biogenesis through phosphorylation of TBCB. Activity is inhibited in cells undergoing apoptosis, potentially due to binding of CDC2L1 and CDC2L2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GHRH
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Ghrh
Gene Accession No. P01286, P09916, P16043
Gene Alias GHRF; Ghrh; GRF; growth hormone releasing hormone; growth hormone-releasing factor; Growth hormone-releasing hormone; INN; MGC119781; OTTHUMP00000030915; Sermorelin; somatocrinin; Somatoliberin; somatorelin
Gene Symbols Ghrh
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14601, 2691, 29446
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.