Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GHRH (Somatoliberin), Sheep anti-Mouse, Rat, Polyclonal, Millipore Sigma™

Catalog No. ABN99325UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN99325UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN99325UL Supplier MilliporeSigma Supplier No. ABN99325UL
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Anti-GHRH (Somatoliberin) Antibody, Cat. No. ABN993, is a highly specific sheep polyclonal antibody that targets Somatoliberin (GHRH) and has been tested in Immunofluorescence, Immunohistochemistry (Paraffin), and Radioimmunoassay.

Somatoliberin (UniProt: P09916; also known as Growth hormone-releasing factor, GRF, Growth hormone-releasing hormone, GHRH) is encoded by the Ghrh gene (Gene ID: 29446) in rat. GHRH F is released by the hypothalamus and acts on the adenohypophyse to stimulate the secretion of growth hormone. The release of growth hormone from pituitary gland depends of the interplay between the stimulatory action of GHRH and inhibitory action of somatotropin release inhibiting factor (SRiF) or somatostatin. These two neurohormones are shown to be released in a reciprocal manner from the median eminence into the portal circulation and result in ultradian rhythmic release pattern of growth hormone.. Post-translational removal of the signal peptide (aa 1-19) and the two propeptide sequences (aa 20-30) and aa 75-104) releases the 43 aa mature Somatoliberin (aa 31-73), which functions as a ligand to stimulate growth hormone (GH) release from the pituitary. Variant receptors for GHRH have been found in several types of tumors, and antagonists of these receptors are shown to block the growth of the tumors. Defects in Ghrh gene are a known to cause of dwarfism, while hypersecretion of the encoded GHRH can lead to gigantism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GHRH (Somatoliberin)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Dilution Radioimmunoassay Analysis: A representative lot detected GHRH (Somatoliberin) in Radioimmunoassay applications (Tannenbaum, G.S., et. al. (1990). Endocrinology. 127(6):3087-95). Immunohistochemistry Analysis: A representative lot detected GHRH (Somatoliberin) in Immunohistochemistry applications (Tannenbaum, G.S., et. al. (1990). Endocrinology. 127(5):2551-60; Epelbaum, J., et. al. (1989). J Neuroendocrinol . 1(2):109-15). Immunofluorescence Analysis: A representative lot detected GHRH (Somatoliberin) in Immunofluorescence applications (Epelbaum, J., et. al. (2009). J Neurosci . 29(25):8198-205).
Formulation Sheep polyclonal antiserum with 0.01% sodium azide.
Gene Alias Somatoliberin;Growth hormone-releasing factor;GRF;Growth hormone-releasing hormone
Gene Symbols Ghrh
Host Species Sheep
Immunogen BSA-conjugated synthetic linear peptide corresponding to the first 29 amino acids from rat Somatoliberin (GHRH).
Molecular Weight of Antigen 12.2 kDa calculated.
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This sheep polyclonal antibody detects somatolierin (GHRH) in mouse and rat brain sections. It targets an epitope within the first 29 amino acids.
Target Species Mouse, Rat
Content And Storage Stable for 1 year at -20°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.