Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GHRHR Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988728
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8728 Supplier Bioss Supplier No. BS8867R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Growth Hormone Releasing Hormone (GHRH) is a member of a family of peptide hormones that also includes VIP (vasoactive intestinal polypeptide) and PACAP (pituitary adenylate cyclase-activating polypeptide). It is a hypothalamic peptide that binds to G-protein coupled receptors to stimulate secretion and synthesis of growth hormone (GH). GHRH receptor proteins have been identified on the plasma membrane of primary human tumor cells, and are thought to be involved in the growth of a variety of cancer cells.

Specifications

Antigen GHRHR
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Ghrhr
Gene Accession No. Q02643
Gene Alias GHRFR; GHRH receptor; Ghrhr; Ghrh-r; GHRHREC; GHRHRpsv; GRF receptor; Grfr; Growth hormone - releasing receptor; growth hormone releasing hormone receptor; Growth hormone-releasing factor receptor; Growth hormone-releasing hormone receptor; IGHD1B; lit; little
Gene Symbols Ghrhr
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GHRHR.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2692
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.