Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GHRL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501728641 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8641 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8641 Supplier Proteintech Group Inc Supplier No. 133091AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ghrelin is a potent stimulator of growth hormone production from the anterior pituitary gland, and is known to play a role in a number of different physiological processes. It is a 28-amino acid peptide existing in two major forms. Ghrelin plays a significant role in neurotrophy, and is essential for cognitive adaptation to changing environments and the process of learning. Ghrelin plays a significant role in various kinase dependent pathways such as AKT in activating the endothelial isoform of nitric oxide synthase.

Specifications

Antigen GHRL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GHRL
Gene Accession No. Q9EQX0, Q9QYH7, Q9UBU3
Gene Alias Appetite regulating hormone, Ghrelin Obestatin, GHRL, Growth hormone secretagogue, Motilin related peptide, MTLRP, obestatin, Protein M46
Gene Symbols GHRL
Host Species Rabbit
Immunogen GHRL Fusion Protein Ag4141
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51738, 58991, 59301
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.