Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GHRPR/Ghrelin Receptor/GHS-R1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981447
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1447 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1447 Supplier Bioss Supplier No. BS11529R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Growth hormone secretagogue receptor 1a (GHSR) is a member of the G-protein coupled receptor family. GHSR may play a role in energy homeostasis and regulation of body weight. Two identified transcript variants are expressed in several tissues and are evolutionary conserved in fish and swine. One transcript, 1a, excises an intron and encodes the functional protein; this protein is the receptor for the Ghrelin ligand and defines a neuroendocrine pathway for growth hormone release. The second transcript (1b) retains the intron and does not function as a receptor for Ghrelin; however, it may function to attenuate activity of isoform 1a. Mutations to the gene can cause isolated partial growth hormone deficiency.

Specifications

Antigen GHRPR/Ghrelin Receptor/GHS-R1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Ghsr
Gene Accession No. Q92847
Gene Alias C530020I22Rik; GHDP; GH-releasing peptide receptor; Ghrelin receptor; GHRP; Ghrp receptor; GHS receptor; GHSR; GHS-R; G-protein coupled receptor; growth hormone secretagogue receptor; Growth hormone secretagogue receptor type 1
Gene Symbols Ghsr
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Ghrelin Receptor.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2693
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.