Learn More
ZING™ Enterprises GHS Introduction Poster
Description
Poster is printed on recycled plastic to support sustainability initiatives and LEED credits, measure 18 in.H x 24 in.W. Made in the USA.
ZING GHS Labeling Poster defines the 6 labeling elements and 9 pictograms used in the Globally Harmonized System. Poster is printed on recycled plastic to support sustainability initiatives and LEED credits, measure 18 in.H x 24 in.W. Made in the USA.
ZING GHS Safety Data Sheet Poster defines the 16 section format used in the Globally Harmonized System. Poster is printed on recycled plastic to support sustainability initiatives and LEED credits, measure 18 in.H x 24 in.W. Made in the USA.
ZING GHS SDS Poster set supports company compliance and training programs related to the new GHS/Hazard Communication requirements. Set includes 3 posters (1) General Introduction Poster: defines the Globally Harmonized System, the 3 major changes and how the changes benefit companies, (2) GHS Labeling Poster: defines the 6 labeling elements and 9 pictograms used in the Globally Harmonized System, (3) GHS Safety Data Sheet Poster defines the 16 section format used in the Globally Harmonized System.
Specifications
Specifications
Material | Plastic |
Quantity | 1 Ea. |
Description | Right to Understand |
Mounting Type | Self-adhesive |
For Use With (Application) | Communication |
Height (English) | 24 in. |
Width (English) | 18 in. |
Languages | English |
Dimensions (L x W) | 24 x 18 in. |
Displayed Text | Hazard Communication |
Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.