Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ GHS Labeled Right-to-Know Safety-Vented Wash Bottles
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Assorted
Deionized Water
Dichloromethane
Ethanol
Ethyl Acetate
Isopropanol
Methanol
Sodium Hypochlorite (Bleach)
Water
Closure Color:
Assorted
Blue
Green
Natural
Red
Yellow
Description
Unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position.
- Pre-printed bottles with chemical name and formula, GHS pictogram and signal word; National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
- Translucent LDPE bottles, Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm
- Wide mouth 53 mm (2 in.) for easy refilling
- Color-coded polypropylene closures aid in quick and easy identification
- Assortment bag includes one each: Acetone, Isopropanol, Bleach and Ethanol
- Bottle with Label: TOLUENE, Red LDPE bottle, does not include vented valve
- Bottle with Label: DEIONIZED H2O, DISTILLED H20 and WATER, does not indicate hazardous chemical and does not have or require GHS pictogram or signal word
- Bottle with Label: METHYL ETHYL KETONE, does not include vented valve
Compliance
Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA’s HazCom 2012 requirements – as pertains to Workplace Labels.
Specifications
Specifications
Capacity | 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16.91 oz. |
Color | Translucent |
Material | LDPE |
Quantity | 3/Pk. |
Sterility | Non-sterile |
Closure Color | Green |
Closure Included | Yes |
Disposable | Reusable |
Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction