Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GI 254023X
GREENER_CHOICESDP

Catalog No. 39951 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-951 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 39-951 Supplier R&D Systems™ Supplier No. 3995/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective ADAM10 metalloprotease inhibitor

Selective ADAM10 metalloprotease inhibitor; displays over 100-fold higher potency at ADAM10 compared to ADAM17. Blocks constitutive release of IL-6R, CX3CL1 and CXCL16 in cell-based cleavage experiments. Inhibits calcium ionophore-induced betacellulin shedding in IMPE cells. Prevents E-cadherin cleavage in A549 cells. Inhibits ADAM10 mediated neuronal outgrowth of dorsal root ganglion neurons in vitro.

Chemical Identifiers

CAS 260264-93-5
Molecular Formula C21H33N3O4

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 391.5
Purity >98%
Molecular Weight (g/mol) 391.51
Chemical Name or Material GI 254023X
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.