Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GI 530159
GREENER_CHOICESDP

Catalog No. 668950 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-895-0 50 mg
66-891-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 66-895-0 Supplier Tocris Bioscience™ Supplier No. 6689/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

TREK1 and TREK2 channel activator

GI 530159 is a TREK1 and TREK2 channel activator (EC50 = 0.76 μM in Rb efflux assay in TREK1-expressing CHO cells). Exhibits selectivity for TREK1/2 over TRAAK, TASK3 and a range of other potassium channels. Hyperpolarizes membrane potential of dorsal root ganglion neurons and depresses neuronal activity in vitro.

Specifications

Quantity 50 mg
Molecular Formula C27H20F6N2O2
Formula Weight 518.45
Product Type Channel Activator
Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.