Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Giardia lamblia Antibody (BB1.1E5), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308870IR
Description
Giardia lamblia Monoclonal antibody specifically detects Giardia lamblia in Protozoa samples. It is validated for ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
Giardia lamblia | |
Monoclonal | |
DyLight 755 | |
G lamblia; Giardia intestinalis | |
Giardia lamblia | |
100 μg | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA, Immunofluorescence | |
BB1.1E5 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Protozoa | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction