Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Giardia lamblia cysts Monoclonal Antibody (I43B), Invitrogen™

Catalog No. MA17441
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17441 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17441 Supplier Thermo Scientific Supplier No. MA17441
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7441 antibody reacts with Giardia lamblia cysts in protozoan samples. MA1-7441 has been successfully used in Immunofluorescence and ELISA applications. The MA1-7441 antibody was raised against purified giardia cysts.

Giardiasisis a diarrhoeal illness caused by a single celled microscopic protozoan parasite, Giardia lamblia, also known as Giardia intestinalis. Giardia lamblia exists in two forms, an active form called a trophozoite, and an inactive form called a cyst. The active trophozoite attaches to the lining of the small intestine and is responsible for causing the signs and symptoms of giardiasis. The trophozoite cannot live long outside of the body and spread of infection is via the cyst which is excreted in the host′s faeces. When it is ingested, stomach acid activates the cyst, and the cyst develops into the disease causing trophozoite in the new host. Giardiasis is diagnosed by finding cysts or trophozoites in the faeces.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Giardia lamblia cysts
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone I43B
Concentration 0.1 mg/mL
Conjugate Unconjugated
Gene Alias G. lamblia; Giardia intestinalis
Host Species Mouse
Immunogen Purified giardia cysts.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.