Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Giardia lamblia cysts Polyclonal Antibody, FITC

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA173151
Description
The PA1-73151 antibody reacts with Giardia lamblia cysts. PA1-73151 has been successfully used in Immunofluorescence applications. The PA1-73151 antibody was raised against purified, intact giardia cysts.
Giardiasisis a diarrhoeal illness caused by a single celled microscopic protozoan parasite, Giardia lamblia, also known as Giardia intestinalis. Giardia lamblia exists in two forms, an active form called a trophozoite, and an inactive form called a cyst. The active trophozoite attaches to the lining of the small intestine and is responsible for causing the signs and symptoms of giardiasis. The trophozoite cannot live long outside of the body and spread of infection is via the cyst which is excreted in the host's faeces. When it is ingested, stomach acid activates the cyst, and the cyst develops into the disease causing trophozoite in the new host. Giardiasis is diagnosed by finding cysts or trophozoites in the faeces.
Specifications
| Giardia lamblia cysts | |
| Polyclonal | |
| FITC | |
| G. lamblia | |
| Purified, intact giardia cysts. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
| Immunocytochemistry | |
| 5 mg/mL | |
| PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
| Goat | |
| IgG fraction | |
| RUO | |
| Protozoa | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction