Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Giardia lamblia Mouse anti-Protozoa, Clone: BB1.1E5, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
| Antigen | Giardia lamblia |
|---|---|
| Clone | BB1.1E5 |
| Concentration | 0.2 mg/ml |
| Dilution | ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml |
| Applications | ELISA, Immunofluorescence |
Description
Giardia lamblia Monoclonal specifically detects Giardia lamblia in Protozoa samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Giardia lamblia | |
| 0.2 mg/ml | |
| ELISA, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Protozoa | |
| G lamblia; Giardia intestinalis | |
| IgG1 κ | |
| Protein A or G purified |
| BB1.1E5 | |
| ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| Giardia lamblia | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title