Learn More
Gibberellic acid, 98%
Description
Gibberellic acid is used as a plant growth hormone. It is also used to trigger germination in dormant seeds in laboratory and green house settings and to stimulate rapid stem and root growth and induce mitotic division in the leaves of some plants. It also serves in grape growing industry as a hormone to induce the production of larger bundles and bigger grape.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 222°C (decomposition) |
| Color | White |
| Quantity | 10 g |
| Beilstein | 54346 |
| Merck Index | 14,4419 |
| Solubility Information | Slightly soluble in water. Soluble in alcohol,isopropyl alcohol,ethanol,acetone and chloroform. |
| Optical Rotation | +78° (c=2 in methanol) |
| Formula Weight | 346.37 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Show More |
Safety and Handling
| Hazard Category | H319 |
| Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
| Precautionary Statement | P264b-P280i-P305+P351+P338 |
| EINECSNumber | 201-001-0 |
| RTECSNumber | LY8990000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.