Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibberellic acid, 98%

Catalog No. AA4556904
Change view
Click to view available options
Quantity:
500 mg
2 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4556904 2 g
AA45569ME 500 mg
AA4556909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4556904 Supplier Thermo Scientific Chemicals Supplier No. 04556904
Only null left
Add to Cart

Plant growth hormone | CAS: 77-06-5 | C19H22O6 | 346.38 g/mol

Gibberellic acid is used as a plant growth hormone. It is also used to trigger germination in dormant seeds in laboratory and green house settings and to stimulate rapid stem and root growth and induce mitotic division in the leaves of some plants. It also serves in grape growing industry as a hormone to induce the production of larger bundles and bigger grape.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-06-5
Molecular Formula C19H22O6
Molecular Weight (g/mol) 346.38
MDL Number MFCD00079329
InChI Key IXORZMNAPKEEDV-QTWFBFKQSA-N
Synonym gibberellic acid, gibberellin a3, gibberellin, gibreskol, brellin, cekugib, grocel, gibberellin x, gibberellic acid ga3, gib-tabs
PubChem CID 91757643
IUPAC Name (1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid
SMILES CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O

Specifications

Melting Point 222°C (decomposition)
Color White
Quantity 2 g
Beilstein 54346
Merck Index 14,4419
Solubility Information Slightly soluble in water. Soluble in alcohol,isopropyl alcohol,ethanol,acetone and chloroform.
Optical Rotation +78° (c=2 in methanol)
Formula Weight 346.37
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Gibberellic acid
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 201-001-0
RTECSNumber LY8990000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.