Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ AmnioMAX™-II Complete Medium
GREENER_CHOICE

Catalog No. 11269016
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-269-016 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-269-016 Supplier Gibco™ Supplier No. 11269016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fully-supplemented medium developed for short term culture of human amniotic fluid cells for cytogenetic studies and in vitro diagnostic procedures

Gibco™ AmnioMAX™ - II Complete Medium is a fully-supplemented medium developed for the short term culture of human amniotic fluid cells for cytogenetic studies and in vitro diagnostic procedures.

Gibco™ AmnioMAX™ - II Complete Medium features:

  • Ready-to-use format
  • Quality and performance testing
  • Unique, optimized formulation

Ready-to-use format

Gibco™ AmnioMAX™ - II Complete Medium is a frozen, 1X medium, ready to use upon thawing. No supplementation is required. Thawed medium can be stored at 2–8°C for up to ten days.

Quality and performance testing

Every lot of Gibco™ AmnioMAX™ - II Complete Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Pooled primary human amniotic fluid samples are cultured for six days in Gibco™ AmnioMAX™ - II Complete Medium before measuring the total number of colonies and total number of mitotic colonies. In addition, each lot is tested for pH and osmolality and must pass a sterility test prior to lot release.

Unique, optimized formulation

Gibco™ AmnioMAX™ - II Complete Medium contains Fetal Bovine Serum (FBS), gentamicin, and L-glutamine to maximize cell attachment and growth. This optimized medium also has an enhanced buffering system that provides greater pH stability during culture manipulations.

Product Use

For in vitro diagnostic use.

cGMP Manufacturing and Quality System

Gibco™ AmnioMAX™ - II Complete Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Order Info

Shipping Condition: Dry Ice

Compliance

Gibco AmnioMAX - II Complete Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Amniocyte Cells
Concentration 1 X
Form Liquid
Product Type Complete Medium
Sterility Sterile-filtered
With Additives Glutamine
Product Line AmnioMAX
Quantity 100 mL
Shipping Condition Dry Ice
Green Features ACT Ecolabel (Silver)
Included Antibiotics Gentamicin
Content And Storage Storage conditions: -5 to -20°C. Protect from light.
Shipping conditions: Frozen
Shelf life: 18 months from date of manufacture if frozen. If stored at 4°C (protected from light): use within 7–10 days once thawed and opened. Extended shelf-life of 50 days if thawed and unopened.
Culture Type Mammalian Cell Culture
Show More Show Less
Can I supplement AmnioMax-II Complete Medium (Cat. No. 11269016) with Fungizone?

No. We do not recommend supplementing AmnioMAX-II Complete Medium (Cat. No. 1129016) with Fungizone, as it can be detrimental to amniocyte growth.  

WARNING: Cancer - www.P65Warnings.ca.gov

In vitro diagnostic

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.