Learn More
Gibco™ DMEM/F-12, GlutaMAX™ supplement


Description
Requires
Supplementation
DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM/F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of DMEM/F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This DMEM / F-12 is modified as follows:
With: GlutaMAX, Phenol Red
Without: HEPES
The complete formulation is available.
Using DMEM/F-12
DMEM/F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with F-12's wide variety of components. DMEM/F-12 with GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. DMEM/F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM/F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM/F-12 uses a sodium bicarbonate buffer system and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature

Specifications
Specifications
Cell Line | MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts |
Classification | Animal Origin-free |
Concentration | 1 X |
Form | Liquid |
Product Type | DMEM (Dulbecco's Modified Eagle Medium)/F-12 |
Serum Level | Standard Serum Supplementation |
Sterility | Sterile-filtered |
Sterilization Method | Sterile-filtered |
With Additives | High Glucose, GlutaMAX, Phenol Red, Sodium Pyruvate |
Without Additives | No HEPES |
Show More |
Safety and Handling
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.