Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ DMEM, high glucose, GlutaMAX™ Supplement, HEPES
GREENER_CHOICE

Catalog No. 10564011
Encompass_Preferred
Click to view available options
Quantity:
500 mL
5 L
10 L
10 x 500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Requires

Supplementation

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells.

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM is modified as follows:

With: High Glucose, HEPES, GlutaMAX, Phenol Red

Without: Sodium Pyruvate

The complete formulation is available.

Using DMEM

DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM with GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line HeLa, 293, Cos-7, and PC-12
Cell Type Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type DMEM (Dulbecco's Modified Eagle Medium)
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives High Glucose, GlutaMAX, HEPES, Phenol Red
Without Additives No Sodium Pyruvate
For Use With (Application) Mammalian cell culture
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco™, GlutaMAX™
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.