Learn More
Gibco™ DPBS, powder, calcium, magnesium

Description
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
This DPBS is modified as follows:
With: Calcium, Magnesium
Without: Phenol Red
Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).
Order Info
Shipping Condition: Room Temperature

Specifications
Specifications
Chemical Name or Material | DPBS |
For Use With (Application) | Mammalian Cell Culture |
pH | 7.3 - 7.5 |
Physical Form | Powder |
Product Line | Gibco™ |
Quantity | 10 x 1 L |
Solution Type | Dulbecco's Phosphate Buffered Saline |
Includes | No Sodium Pyruvate |
Manufacturing Quality | cGMP-compliant under the ISO 13485 standard |
Osmolality | 260 - 310 mOsm/kg |
For in vitro diagnostic use. Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.