Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ DPBS, no calcium, no magnesium
GREENER_CHOICE

Catalog No. 14190136
Encompass_Preferred
Click to view available options
Quantity:
500 mL
1000 mL
5 L
10 L
10 x 500 mL
20 L
6 x 1000 mL
Packaging:
Bag
Bottle

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

This DPBS is modified as follows:

Without: Calcium, Magnesium, Phenol Red

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DPBS
Color Clear
Concentration 1X
For Use With (Application) Mammalian Cell Culture
pH 7.0 to 7.3
Physical Form Liquid
Product Line Gibco™
Quantity 1000 mL
Solution Type Dulbecco's Phosphate Buffered Saline
Packaging Bottle
Sterility Sterile-filtered
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 270 - 300 mOsm/kg
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.