Learn More
Gibco™ DPBS, no calcium, no magnesium

Description
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.
This DPBS is modified as follows:
Without: Calcium, Magnesium, Phenol Red
Order Info
Shipping Condition: Room Temperature

Specifications
Specifications
Chemical Name or Material | DPBS |
Concentration | 1 X |
For Use With (Application) | Mammalian Cell Culture |
pH | 7.0 to 7.3 |
Physical Form | Liquid |
Product Line | Gibco™ |
Quantity | 20 L |
Solution Type | Dulbecco's Phosphate Buffered Saline |
Packaging | Bag |
Sterility | Sterile-filtered |
Show More |
Safety and Handling
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.