Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Fetal Bovine Serum, certified, heat inactivated, United States

Catalog No. A3840002
Encompass_Preferred
Click to view available options
Quantity:
500 mL
10 x 50 mL
Format:
Bottle
One Shot

Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera Category: Premium (Performance Plus)
  • Origin: United States
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤15 mg/dL

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Description Fetal Bovine Serum, heat inactivated, certified, One Shot™, United States
Species Bovine
Age Fetal
Country of Origin United States
Form Liquid
Format One Shot
Product Type Fetal Bovine Serum
Quantity 10 x 50 mL
Serum Treatment Heat-inactivated
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Processing Triple Filtered at 0.1 μm
Endotoxin Concentration ≤5 EU/mL
Heat Inactivated Yes
Hemoglobin Concentration ≤15 mg/dL
Purity or Quality Grade Certified
Product Line One Shot™
Shipping Condition Frozen
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.