Learn More
Gibco™ Embryonic stem-cell FBS, qualified, USDA-approved regions

Description
Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera Category: Specialty
- Origin: Countries meeting USDA importation requirements
- Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
- Hemoglobin level: ≤25 mg/dL
Specially tested for the ability to sustain undifferentiated cellular morphology of embryonic stem cells.
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
| Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
| Species | Cattle/Bovine |
| Age | Fetal |
| Country of Origin | Mexico |
| Form | Liquid |
| Format | One Shot |
| Product Type | Fetal Bovine Serum |
| Quantity | 50 mL |
| Serum Treatment | Standard (Sterile-filtered) |
| Sterility | Sterile |
| Show More |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.