Learn More
Gibco™ Fetal Bovine Serum, qualified, heat inactivated, USDA-approved regions
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
- Replacing with Fetal Bovine Serum, value
- Sera application: for cell culture with robust cell lines, good value for basic research
- Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
- Hemoglobin level: ≤25 mg/dL
- Origin: countries meeting USDA importation requirements
Gibco serum delivers
- ISO 13485 certified, processed in FDA registered facilities
- Triple filtered at 0.1 μm
- Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
| Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
| Species | Cattle/Bovine |
| Age | Fetal |
| Country of Origin | Countries meeting USDA importation requirements |
| Form | Liquid |
| Format | Bottle |
| Product Type | Fetal Bovine Serum |
| Quantity | 500 mL |
| Serum Treatment | Heat-inactivated |
| Sterility | Sterile |
| Show More |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.