Learn More
Gibco™ Human FGF-basic (FGF-2/bFGF) (aa 10-155) Recombinant Protein

Description
Carrier-Free Reconstitution: Centrifuge the vial briefly, before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in sterile, distilled water to 0.1-0.5 mg/mL. Apportion the reconstituted protein into working aliquots and store at ≤-20°C. Make any further dilutions of the reconstituted protein in medium or buffered solution containing carrier protein, such as PBS with 0.1% BSA. Storage: Store the lyophilized protein at 2-8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤-20°C (not in a frost-free freezer). Avoid repeated freeze-thaw cycles.

Specifications
Specifications
Accession Number | P09038 |
For Use With (Application) | Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot |
Formulation | Protein with no preservative |
Gene ID (Entrez) | 2247 |
Molecular Weight (g/mol) | 17 kDa |
Name | Human FGF-basic (FGF-2/bFGF) (aa 10-155) |
Purification Method | Purified |
Quantity | 100 μg |
Storage Requirements | 4°C |
Regulatory Status | RUO |
Show More |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.