GIBCO™ Blasticidin S HCl (10mg/mL), Sterile Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Selective antibiotic in concentration range of 50G–100µg/mL

Manufacturer: Gibco A1113903

 View more versions of this product

Catalog No. A1113903

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Antibiotic Spectrum Blasticidin
    Quantity 10 x 1mL,10 x 1mL
    Regulatory Status For Research Use Only. Not for use in diagnostic procedures.
    Concentration 10mg/mL,10mg/mL
    Solution Type Antibiotic (Selective)
    For Use With (Application) Bacterial Selection;Eukaryotic Selection/Stable Cell Line Generation
    Product Type Blasticidin
    Format Liquid,Liquid

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit