Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Goat Serum, New Zealand origin

Click to view available options
Quantity:
100 mL
500 mL
Description
- Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays
- It has earned the trust of researchers with consistent quality and award-winning support
- Helps meet research needs and budget requirements
- Shipped frozen, store at <-10°C
- Sera Category: Specialty
- Origin: New Zealand
- Age: Adult
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
| Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
| Species | Goat/Caprine |
| Age | Adult |
| Country of Origin | New Zealand |
| Form | Liquid |
| Product Type | Goat Serum |
| Quantity | 500 mL |
| Serum Treatment | Standard (Sterile-filtered) |
| Sterility | Sterile |
| Product Line | Gibco™ |
| Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction