Learn More
Gibco™ Grace's Insect Medium, supplemented

Description
Requires
Grace's Unsupplemented Insect Medium requires ambient CO2 in order to maintain physiological pH.
Originally designed for the growth of cells from the Australian Emperor Gum Moth, Antherea eucalypti. Sf9 and Sf21 cells are routinely used to express recombinant protein using the Baculovirus Expression Vector System (BEVS).
- Contains lactalbumin hydrolysate, yeastolate, L-glutamine, and sodium bicarbonate
- Dual site cGMP manufacturing and quality system
- Commonly supplemented with 10% Fetal Bovine Serum (FBS) before use
Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Cell Line | Sf21, Sf9 |
| Cell Type | Insect Cell |
| Form | Liquid |
| Product Type | Grace's Insect Medium |
| With Additives | Glutamine, Yeastolate, Lactalbumin Hydrolysate |
| Without Additives | No Insect Hemolymph |
| Product Line | Gibco |
| Quantity | 10 x 500 mL |
| Shipping Condition | Room Temperature |
| Inorganic Salts | Calcium, Magnesium |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.