Learn More
Gibco™ HBSS, no calcium, no magnesium


Description
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.
We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.
This HBSS is modified as follows:
With: Phenol Red, Glucose
Without: Calcium, Magnesium
The complete formulation is available.
Order Info
Shipping Condition: Room Temperature

Specifications
Specifications
Additive | No Sodium Pyruvate |
Classification | Animal Origin-Free |
Content And Storage | Storage conditions: 15-30° C Shipping conditions: Ambient Shelf life: 36 months from date of manufacture |
Dilution | 1 X |
Product Line | Gibco™ |
Product Type | Hanks' Balanced Salt Solution (HBSS) |
Quantity | 500 mL |
Solution Type | Hank's Balanced Salt |
For Use With (Application) | Mammalian Cell Culture |
Osmolality | 235 - 295 mOsm/kg |
Show More |
Safety and Handling
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.