Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Hybridoma-SFM
GREENER_CHOICE

Catalog No. p-4924977
Encompass_Preferred
Click to view available options
Quantity:
500 mL
1000 mL

Catalog No. 12-045-084

Supplier: Gibco™ 12045084

Only null left
Add to Cart
Edge
Add to Cart

Specifically developed to support the growth of hybridomas for antibody production

  • Hybridoma-SFM is a serum-free, very low-protein (<20 μg/mL) medium specifically developed to support the growth of hybridomas for antibody production
  • Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas, Hybridoma-SFM provides high cell densities and IgG production
  • Cholesterol-dependent cultures such as NS0 require the addition of 250X Cholesterol Lipid Concentrate
  • Hybridoma-SFM is a complete medium that is ready to use, no supplementation required
  • Store at 2°C to 8°C, protected from light

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Hybridomas
Classification Low Protein, Serum-free
Form Liquid
Product Type Serum Free Medium (SFM)
Serum Level Serum-free
With Additives Glutamine, Phenol Red
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.