Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ IMDM, GlutaMAX™ Supplement
GREENER_CHOICE

Catalog No. 31980097
Encompass_Preferred
Click to view available options
Quantity:
500 mL
10 x 500 mL
This item is not returnable. View return policy

Catalog No. 31-980-097

Supplier: Gibco™ 31980097

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Requires

Supplementation

IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells.

IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This IMDM is modified as follows:

With: GlutaMAX™, Phenol Red

Without: α-thioglycerol, 2-mercaptoethanol

The complete formulation is available.

Using IMDM

IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate. IMDM with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Jurkat, COS-7, and macrophage cells
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type IMDM (Iscove's Modified Dulbecco's Medium)
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives High Glucose, GlutaMAX, HEPES, Phenol Red, Sodium Pyruvate
Without Additives No α-Thioglycerol, No 2-Mercaptoethanol
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco, GlutaMAX
Quantity 10 x 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.