Learn More
Leibovitz's L-15 Medium Non-distribution product as customer accommodation.
Supportsgrowth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue
Manufacturer:  Gibco 11415114
 View more versions of this product
Catalog No. 11-415-114
  Check Availability
Add to cart
Disclaimers:
Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. For in vitro diagnostic use.
Requires : Supplementation
Specifications Description & Specifications
Specifications
Liquid
Sustainable packaging
Storage: 2-8° C. Protect from light. Shelf life: 12 months from date of manufacture
Calcium Chloride (CaCl2) (anhyd.), Choline chloride, D+ Galactose, D-Calcium pantothenate, Folic Acid, Glycine, L-Alanine, L-Arginine, L-Asparagine, L-Cysteine, L-Glutamine, L-Histidine, L-Isoleucine, L-Leucine, L-Lysine, L-Methionine, L-Phenylalanine, L-Serine, L-Threonine, L-Tryptophan, L-Tyrosine, L-Valine, Magnesium Chloride (anhydrous), Magnesium Sulfate (MgSO4) (anhyd.), Niacinamide, Phenol Red, Potassium Chloride (KCl), Potassium Phosphate monobasic (KH2PO4), Pyridoxine hydrochloride, Riboflavin 5-phosphate Na, Sodium Chloride (NaCl), Sodium Phosphate dibasic (Na2HPO4) anhydrous, Sodium Pyruvate, Thiamine monophosphate, i-Inositol
10 x 500mL
Sodium Pyruvate; L-Glutamine; Phenol Red