Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ LV-MAX™ Production Medium

Catalog No. p-7113083
Encompass_Preferred
Click to view available options
Quantity:
1 L
6 x 1 L

LV-MAX Production Medium is a chemically defined, serum-free, protein-free medium, specially developed for high-density growth and transfection of Gibco Viral Production Cells (suspension-adapted HEK 293-derived cells).

Gibco LV-MAX Production Medium is a chemically defined, serum-free, protein-free medium, specially developed for high-density growth and transfection of Gibco Viral Production Cells (suspension-adapted HEK 293-derived cells) promoting high-titer lentiviral production.

  • Supports growth of suspension Viral Production Cells to densities of over 1.0 x 107 cells/mL
  • Enables sustained, high-level production of high-density transiently transfected cultures, achieving titer of up to 1 x 108 TU/mL (unconcentrated LVVs-GFP)
  • Supports growth and transfection of Viral Production Cells in culture formats from 96-deep well blocks to 2-L flasks and bioreactors

LV-MAC Production Medium is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. The medium is not recommended for adherent cell cultures.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Viral Production Cells
Cell Type HEK 293
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Liquid
Product Type Lentiviral Production Medium
Sterility Sterile-filtered
With Additives GlutaMAX
Manufacturing Quality ISO 13485, MDSAP, FDA-registered
Product Line LV-MAX™
Quantity 1 L
Shipping Condition Room Temperature
Included Antibiotics No antibiotics
Content And Storage 1 L bottle; Reagents should be stored at 2°C to 8°C, protected from light
Culture Type Suspension Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.