Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ MEM, NEAA, no glutamine
GREENER_CHOICE

Catalog No. 10370088
Encompass_Preferred
Click to view available options
Quantity:
500 mL
10 x 500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media.

Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This MEM is modified as follows:

With: NEAA, Phenol Red

Without: L-glutamine, HEPES

The complete formulation is available.

Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow's MEM, MEM α, DMEM, and Temin's Modification. MEM is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This product is made with Earle's salts.

MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Cell Type Primary Rat Astrocytes
Classification Animal Origin-free
Concentration 1 X
Product Type MEM (Minimum Essential Medium)
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Low Glucose, Phenol Red, Non Essential Amino Acids (NEAA)
Without Additives No Glutamine, No HEPES, No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco™
Quantity 10 x 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2°C to 8°C (protect from light)
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.