Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ RPMI 1640 Medium, GlutaMAX™ Supplement, HEPES

Catalog No. p-4919941
Encompass_Preferred
Click to view available options
Quantity:
5 L
10 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Requires

Supplementation

Originally developed to culture human leukemic cells in suspension and as a monolayer

RPMI 1640 Medium (Roswell Park Memorial Institute 1640 medium) has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas.

  • Modified as follows: with GlutaMAX™, phenol red, HEPES
  • Contains reducing agent glutathione and high concentrations of vitamins
  • Contains biotin, vitamin B12 and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium
  • Vitamins inositol and choline are present in very high concentrations
  • GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format
  • Contains no proteins, lipids, or growth factors
  • Requires supplementation, commonly with 10% Fetal Bovine Serum (FBS)
  • Uses sodium bicarbonate buffer system (2.0g/L), and therefore requires a 5-10% CO2 environment to maintain physiological pH

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
With Additives GlutaMAX, Phenol Red
Without Additives No Sodium Pyruvate
Quantity 10 L
Formulation Biotin, Calcium nitrate (Ca(NO3)2 4H2O), Choline chloride, D-Calcium pantothenate, D-Glucose (Dextrose), Folic Acid, Glutathione (reduced), Glycine, HEPES, L-Alanyl-Glutamine, L-Arginine, L-Asparagine, L-Aspartic acid, L-Cystine, L-Glutamic Acid, L-Histidine, L-Hydroxyproline, L-Isoleucine, L-Leucine, L-Lysine hydrochloride, L-Phenylalanine, L-Proline, L-Serine, L-Threonine, L-Tryptophan, L-Tyrosine, L-Valine, Magnesium Sulfate (MgSO4-7H2O), Niacinamide, Para-Aminobenzoic Acid, Phenol Red, Potassium Chloride (KCl), Pyridoxine hydrochloride, Riboflavin, Sodium Bicarbonate (NaHCO3), Sodium Chloride (NaCl), Sodium Phosphate dibasic (Na2HPO4) anhydrous, Thiamine hydrochloride, Vitamin B12, i-Inositol
Content And Storage Storage: 2-8° C. Protect from light. Shelf life: 12 months from date of manufacture
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.