Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Schneider's Drosophila Medium
GREENER_CHOICE

Catalog No. p-4924981
Encompass_Preferred
Click to view available options
Quantity:
500 mL
10 x 500 mL

Modified to contain L-glutamine and sodium bicarbonate

Schneider's Drosophila Medium was originally designed for the growth of S2 cells from the fruit fly, Drosophila melanogaser. Schneider's Drosophila Medium can also be used to culture other dipteran cell lines. This medium can be used for transient or stable transfection and large scale protein production in S2 cells.

Formulation

  • Contains unique components required for S2 cell culture, including malic acid, fumaric acid, alpha-ketoglutaric acid, succinic acid, and the sugar trehelos
  • Commonly supplemented with 10% fetal bovine serum (FBS) before use
  • Requires ambient CO2 in order to maintain physiological pH

Cell Culture, Drosophila S2 Cell Culture, Insect Cell Culture

Compliance

Gibco Schneider's Drosophila Medium is manufactured at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line S2 (Schneider 2)
Cell Type Insect Cell
Form Liquid
Product Type Schneider's Drosophila Medium
With Additives Glutamine
Without Additives No HEPES, No Phenol Red, No Sodium Pyruvate
Product Line Gibco™
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.