Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Sf-900™ II SFM
GREENER_CHOICE

Catalog No. p-4924984
Encompass_Preferred
Click to view available options
Quantity:
500 mL
1000 mL
6 x 1 L

Optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells

Sf-900 II SFM is a serum-free, protein-free insect cell culture medium for large-scale production of recombinant proteins expressed using the baculovirus expression vector system (BEVS). This medium is suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines.

  • Superior long-term, high-density growth
  • Optimized for recombinant protein production
  • Serum-free, protein-free, ready-to-use formulation
  • Scalable in bioreactors

Bioproduction, Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture, Viral Vaccine Production

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Sf21, Sf9
Cell Type Insect Cell
Classification Protein-free, Serum-free
Form Liquid
Product Type Insect Cell Serum Free Medium (SFM)
Serum Level Serum-free
With Additives Glutamine
Product Line Gibco™, Sf-900™
Quantity 500 mL
Shipping Condition Room Temperature
Species S. frugiperda, Spodoptera frugiperda
Content And Storage Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.